2'-(Trifluormethyl)biphenyl-3-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid
014253-100MG
014253-1G
FLUO014253-100MGEA
0
EUR
FLUO014253-100MG
FLUO014253-1G
2'-(Trifluormethyl)biphenyl-3-carbonsäure
2'-(Trifluormethyl)biphenyl-3-carbonsäure
Formel:
C₁₄H₉F₃O₂ Molecular Weight: 266,22 g/mol |
MDL:
MFCD03424602 CAS-Nummer: 168618-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...