3'-(Trifluormethyl)biphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

014254-100MG 014254-1G
FLUO014254-100MGEA 0 EUR
FLUO014254-100MG FLUO014254-1G
3'-(Trifluormethyl)biphenyl-3-carbonsäure
3'-(Trifluormethyl)biphenyl-3-carbonsäure
Formel: C₁₄H₉F₃O₂
Molecular Weight: 266,22 g/mol
MDL: MFCD03424603
CAS-Nummer: 168619-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR