4'-(Trifluormethyl)biphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

014255-100MG 014255-1G
FLUO014255-100MGEA 0 EUR
FLUO014255-100MG FLUO014255-1G
4'-(Trifluormethyl)biphenyl-3-carbonsäure
4'-(Trifluormethyl)biphenyl-3-carbonsäure
Formel: C₁₄H₉F₃O₂
Molecular Weight: 266.22 g/mol
MDL: MFCD03424604
CAS-Nummer: 199528-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR