4'-(Trifluormethyl)biphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4856-5G
CA-4856-1G
COBBCA-4856-5GEA
0
EUR
COBBCA-4856-5G
COBBCA-4856-1G
4'-(Trifluormethyl)biphenyl-3-carbonsäure
4'-(Trifluormethyl)biphenyl-3-carbonsäure
Formel:
C₁₄H₉F₃O₂ Molecular Weight: 266.22 g/mol |
MDL:
MFCD03424604 CAS-Nummer: 199528-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...