3'-(Trifluormethyl)biphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 3'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

014258-100MG 014258-1G
FLUO014258-100MGEA 0 EUR
FLUO014258-100MG FLUO014258-1G
3'-(Trifluormethyl)biphenyl-4-carbonsäure
3'-(Trifluormethyl)biphenyl-4-carbonsäure
Formel: C₁₄H₉F₃O₂
Molecular Weight: 266,22 g/mol
MDL: MFCD03424607
CAS-Nummer: 195457-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR