Methyl-4'-brombiphenyl-4-carboxylat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 4'-bromo[1,1'-biphenyl]-4-carboxylate, Methyl 4'-bromo-[1,1'-biphenyl]-4-carboxylate

2W-0305.500 2W-0305.1 2W-0305.5 2W-0305.10
BIOT2W-0305.500EA 0 EUR
BIOT2W-0305.500 BIOT2W-0305.1 BIOT2W-0305.5 BIOT2W-0305.10
Methyl-4'-brombiphenyl-4-carboxylat
Methyl-4'-brombiphenyl-4-carboxylat
Formel: C₁₄H₁₁BrO₂
Molecular Weight: 291.14 g/mol
MDL: MFCD00496638
CAS-Nummer: 89901-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR