2'-Brom-6'-methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4907-1G CA-4907-250M
COBBCA-4907-1GEA 884 EUR
COBBCA-4907-1G COBBCA-4907-250M
2'-Brom-6'-methylbiphenyl-3-carbonsäure
2'-Brom-6'-methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₁BrO₂
Molecular Weight: 291.14 g/mol
MDL: MFCD15143507
CAS-Nummer: 1215206-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR