2'-Brombiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4829-5G CA-4829-1G
COBBCA-4829-5GEA 2460 EUR
COBBCA-4829-5G COBBCA-4829-1G
2'-Brombiphenyl-3-carbonsäure
2'-Brombiphenyl-3-carbonsäure
Formel: C₁₃H₉BrO₂
MDL: MFCD11574776
CAS-Nummer: 1049143-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR