3'-Brombiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4828-5G CA-4828-1G
COBBCA-4828-5GEA 396 EUR
COBBCA-4828-5G COBBCA-4828-1G
3'-Brombiphenyl-3-carbonsäure
3'-Brombiphenyl-3-carbonsäure
Formel: C₁₃H₉BrO₂
Molecular Weight: 277,12 g/mol
MDL: MFCD00093210
CAS-Nummer: 854237-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR