4'-Brom-3'-methoxybiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4929-5G
CA-4929-1G
COBBCA-4929-5GEA
3040
EUR
COBBCA-4929-5G
COBBCA-4929-1G
4'-Brom-3'-methoxybiphenyl-3-carbonsäure
4'-Brom-3'-methoxybiphenyl-3-carbonsäure
Formel:
C₁₄H₁₁BrO₃ Molecular Weight: 307.14 g/mol |
MDL:
MFCD15143523 CAS-Nummer: 1215206-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...