3'-Chlorbiphenyl-2-carbonsäure

Lieferant: FLUOROCHEM

011486-500MG 011486-1G
FLUO011486-500MGEA 0 EUR
FLUO011486-500MG FLUO011486-1G
3'-Chlorbiphenyl-2-carbonsäure
3'-Chlorbiphenyl-2-carbonsäure
Formel: C₁₃H₉ClO₂
CAS-Nummer: 73178-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR