2',4'-Dichlorbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4919-1G CA-4919-250M
COBBCA-4919-1GEA 742 EUR
COBBCA-4919-1G COBBCA-4919-250M
2',4'-Dichlorbiphenyl-3-carbonsäure
2',4'-Dichlorbiphenyl-3-carbonsäure
Formel: C₁₃H₈Cl₂O₂
MDL: MFCD11574827
CAS-Nummer: 380228-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR