2',5'-Dichlorbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4917-1G CA-4917-250M
COBBCA-4917-1GEA 742 EUR
COBBCA-4917-1G COBBCA-4917-250M
2',5'-Dichlorbiphenyl-3-carbonsäure
2',5'-Dichlorbiphenyl-3-carbonsäure
Formel: C₁₃H₈Cl₂O₂
MDL: MFCD11574811
CAS-Nummer: 1049036-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR