2'-Chlorbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4825-1G CA-4825-5G
COBBCA-4825-1GEA 188 EUR
COBBCA-4825-1G COBBCA-4825-5G
2'-Chlorbiphenyl-3-carbonsäure
2'-Chlorbiphenyl-3-carbonsäure
Formel: C₁₃H₉ClO₂
MDL: MFCD03424596
CAS-Nummer: 168619-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR