3'-Chlorbiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4826-5G
CA-4826-1G
COBBCA-4826-5GEA
549
EUR
COBBCA-4826-5G
COBBCA-4826-1G
3'-Chlorbiphenyl-3-carbonsäure
3'-Chlorbiphenyl-3-carbonsäure
Formel:
C₁₃H₉ClO₂ Molecular Weight: 232,67 g/mol |
MDL:
MFCD03424595 CAS-Nummer: 168619-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...