4'-Chlorbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4'-Chloro-[1,1'-biphenyl]-3-carboxylic acid

CA-4827-1G CA-4827-5G
COBBCA-4827-1GEA 183 EUR
COBBCA-4827-1G COBBCA-4827-5G
4'-Chlorbiphenyl-3-carbonsäure
4'-Chlorbiphenyl-3-carbonsäure
Formel: C₁₃H₉ClO₂
Molecular Weight: 232,67 g/mol
MDL: MFCD03426505
CAS-Nummer: 4655-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR