4'-Chlorbiphenyl-3-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
4'-Chloro-[1,1'-biphenyl]-3-carboxylic acid
011497-500MG
011497-1G
FLUO011497-500MGEA
0
EUR
FLUO011497-500MG
FLUO011497-1G
4'-Chlorbiphenyl-3-carbonsäure
4'-Chlorbiphenyl-3-carbonsäure
Formel:
C₁₃H₉ClO₂ Molecular Weight: 232,67 g/mol |
MDL:
MFCD03426505 CAS-Nummer: 4655-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...