4'-Chlorbiphenyl-4-carbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 4'-Chloro[1,1'-biphenyl]-4-carboxylic acid, 4'-Chloro-[1,1'-biphenyl]-4-carboxylic acid

2X-0702.500 2X-0702.1 2X-0702.5 2X-0702.10
BIOT2X-0702.500EA 0 EUR
BIOT2X-0702.500 BIOT2X-0702.1 BIOT2X-0702.5 BIOT2X-0702.10
4'-Chlorbiphenyl-4-carbonsäure
4'-Chlorbiphenyl-4-carbonsäure
Formel: C₁₃H₉ClO₂
MDL: MFCD03424592
CAS-Nummer: 5748-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR