3'-Chlor-4'-methoxybiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4946-5G
CA-4946-1G
COBBCA-4946-5GEA
3040
EUR
COBBCA-4946-5G
COBBCA-4946-1G
3'-Chlor-4'-methoxybiphenyl-3-carbonsäure
3'-Chlor-4'-methoxybiphenyl-3-carbonsäure
Formel:
C₁₄H₁₁ClO₃ Molecular Weight: 262.69 g/mol |
MDL:
MFCD11932292 CAS-Nummer: 1181237-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...