3'-Chlor-5'-methoxybiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4926-1G
CA-4926-5G
COBBCA-4926-1GEA
872
EUR
COBBCA-4926-1G
COBBCA-4926-5G
3'-Chlor-5'-methoxybiphenyl-3-carbonsäure
3'-Chlor-5'-methoxybiphenyl-3-carbonsäure
Formel:
C₁₄H₁₁ClO₃ Molecular Weight: 262.69 g/mol |
MDL:
MFCD15143520 CAS-Nummer: 1215206-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...