4'-Chlor-3'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4925-5G CA-4925-1G
COBBCA-4925-5GEA 3080 EUR
COBBCA-4925-5G COBBCA-4925-1G
4'-Chlor-3'-methoxybiphenyl-3-carbonsäure
4'-Chlor-3'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁ClO₃
Molecular Weight: 262.69 g/mol
MDL: MFCD15143519
CAS-Nummer: 1215205-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR