4'-Chlor-3'-fluorbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4855-5G CA-4855-1G
COBBCA-4855-5GEA 2460 EUR
COBBCA-4855-5G COBBCA-4855-1G
4'-Chlor-3'-fluorbiphenyl-3-carbonsäure
4'-Chlor-3'-fluorbiphenyl-3-carbonsäure
Formel: C₁₃H₈ClFO₂
MDL: MFCD06201433
CAS-Nummer: 844878-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR