2'-Cyanbiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4863-1G
CA-4863-5G
COBBCA-4863-1GEA
451
EUR
COBBCA-4863-1G
COBBCA-4863-5G
2'-Cyanbiphenyl-3-carbonsäure
2'-Cyanbiphenyl-3-carbonsäure
Formel:
C₁₄H₉NO₂ |
MDL:
MFCD06802573 CAS-Nummer: 893736-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...