3'-Cyanbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4865-5G CA-4865-1G
COBBCA-4865-5GEA 1370 EUR
COBBCA-4865-5G COBBCA-4865-1G
3'-Cyanbiphenyl-3-carbonsäure
3'-Cyanbiphenyl-3-carbonsäure
Formel: C₁₄H₉NO₂
Molecular Weight: 223.23 g/mol
MDL: MFCD04117360
CAS-Nummer: 253878-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR