4'-Cyanbiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4864-1G
CA-4864-5G
COBBCA-4864-1GEA
124
EUR
COBBCA-4864-1G
COBBCA-4864-5G
4'-Cyanbiphenyl-3-carbonsäure
4'-Cyanbiphenyl-3-carbonsäure
Formel:
C₁₄H₉NO₂ Molecular Weight: 223.23 g/mol |
MDL:
MFCD04117352 CAS-Nummer: 149506-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...