4'-Cyanbiphenyl-4-carbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 4'-Cyano[1,1'-biphenyl]-4-carboxylic acid, 4'-Cyano-[1,1'-biphenyl]-4-carboxylic acid

12N-132.500 12N-132.1 12N-132.5 12N-132.10
BIOT12N-132.500EA 0 EUR
BIOT12N-132.500 BIOT12N-132.1 BIOT12N-132.5 BIOT12N-132.10
4'-Cyanbiphenyl-4-carbonsäure
4'-Cyanbiphenyl-4-carbonsäure
Formel: C₁₄H₉NO₂
Molecular Weight: 223,23 g/mol
MDL: MFCD00438676
CAS-Nummer: 5728-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR