2'-(Dimethylamino)biphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4882-1G CA-4882-250M
COBBCA-4882-1GEA 864 EUR
COBBCA-4882-1G COBBCA-4882-250M
2'-(Dimethylamino)biphenyl-3-carbonsäure
2'-(Dimethylamino)biphenyl-3-carbonsäure
Formel: C₁₅H₁₅NO₂
MDL: MFCD15143494
CAS-Nummer: 1175653-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR