3'-(Dimethylamino)biphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4883-1G CA-4883-250M
COBBCA-4883-1GEA 0 EUR
COBBCA-4883-1G COBBCA-4883-250M
3'-(Dimethylamino)biphenyl-3-carbonsäure
3'-(Dimethylamino)biphenyl-3-carbonsäure
Formel: C₁₅H₁₅NO₂
MDL: MFCD11895576
CAS-Nummer: 1181320-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR