4'-(Dimethylamino)biphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4884-5G CA-4884-1G CA-4884-250M
COBBCA-4884-5GEA 2350 EUR
COBBCA-4884-5G COBBCA-4884-1G COBBCA-4884-250M
4'-(Dimethylamino)biphenyl-3-carbonsäure
4'-(Dimethylamino)biphenyl-3-carbonsäure
Formel: C₁₅H₁₅NO₂
MDL: MFCD06802623
CAS-Nummer: 893737-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR