3'-Aminobiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3'-Amino-[1,1'-biphenyl]-3-carboxylic acid

CA-4866-5G CA-4866-1G
COBBCA-4866-5GEA 1090 EUR
COBBCA-4866-5G COBBCA-4866-1G
3'-Aminobiphenyl-3-carbonsäure
3'-Aminobiphenyl-3-carbonsäure
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213,24 g/mol
MDL: MFCD03839954
CAS-Nummer: 124221-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR