4'-Aminobiphenyl-4-carbonsäure 97%

Lieferant: COMBI-BLOCKS
Achtung

CA-4868-1G CA-4868-250M
COBBCA-4868-1GEA 254 EUR
COBBCA-4868-1G COBBCA-4868-250M
4'-Aminobiphenyl-4-carbonsäure 97%
4'-Aminobiphenyl-4-carbonsäure
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03990480
CAS-Nummer: 85096-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR