3'-Nitrobiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4835-5G CA-4835-1G
COBBCA-4835-5GEA 2150 EUR
COBBCA-4835-5G COBBCA-4835-1G
3'-Nitrobiphenyl-3-carbonsäure
3'-Nitrobiphenyl-3-carbonsäure
Formel: C₁₃H₉NO₄
MDL: MFCD06803016
CAS-Nummer: 149505-87-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR