4'-Nitrobiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4834-5G CA-4834-1G
COBBCA-4834-5GEA 2510 EUR
COBBCA-4834-5G COBBCA-4834-1G
4'-Nitrobiphenyl-3-carbonsäure
4'-Nitrobiphenyl-3-carbonsäure
Formel: C₁₃H₉NO₄
MDL: MFCD11895693
CAS-Nummer: 729-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR