4'-Nitrobiphenyl-4-carbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 4'-Nitro-[1,1'-biphenyl]-4-carboxylic acid, 4'-Nitro[1,1'-biphenyl]-4-carboxylic acid

3X-0716.500 3X-0716.1 3X-0716.5 3X-0716.10
BIOT3X-0716.500EA 0 EUR
BIOT3X-0716.500 BIOT3X-0716.1 BIOT3X-0716.5 BIOT3X-0716.10
4'-Nitrobiphenyl-4-carbonsäure
4'-Nitrobiphenyl-4-carbonsäure
Formel: C₁₃H₉NO₄
Molecular Weight: 243.22 g/mol
MDL: MFCD00043911
CAS-Nummer: 92-89-7
EINECS: 202-201-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR