2'-Methoxy-6'-methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4912-1G CA-4912-250M
COBBCA-4912-1GEA 864 EUR
COBBCA-4912-1G COBBCA-4912-250M
2'-Methoxy-6'-methylbiphenyl-3-carbonsäure
2'-Methoxy-6'-methylbiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₃
MDL: MFCD15143511
CAS-Nummer: 1215206-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR