2'-Methoxybiphenyl-3-carbonsäure
Lieferant: FLUOROCHEM
014246-100MG
014246-1G
FLUO014246-100MGEA
0
EUR
FLUO014246-100MG
FLUO014246-1G
2'-Methoxybiphenyl-3-carbonsäure
2'-Methoxybiphenyl-3-carbonsäure
Formel:
C₁₄H₁₂O₃ Molecular Weight: 228.25 g/mol |
MDL:
MFCD03424597 CAS-Nummer: 168618-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...