3'-Methoxy-5'-methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4922-1G CA-4922-5G
COBBCA-4922-1GEA 623 EUR
COBBCA-4922-1G COBBCA-4922-5G
3'-Methoxy-5'-methylbiphenyl-3-carbonsäure
3'-Methoxy-5'-methylbiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₃
MDL: MFCD15143516
CAS-Nummer: 1215205-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR