4'-Methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4821-5G CA-4821-1G
COBBCA-4821-5GEA 174 EUR
COBBCA-4821-5G COBBCA-4821-1G
4'-Methoxybiphenyl-3-carbonsäure
4'-Methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
MDL: MFCD03424599
CAS-Nummer: 725-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR