2'-Methoxybiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 2'-Methoxy-[1,1'-biphenyl]-4-carboxylic acid

014249-100MG 014249-1G
FLUO014249-100MGEA 0 EUR
FLUO014249-100MG FLUO014249-1G
2'-Methoxybiphenyl-4-carbonsäure
2'-Methoxybiphenyl-4-carbonsäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
MDL: MFCD03424600
CAS-Nummer: 5728-32-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR