3'-Methoxybiphenyl-4-carbonsäure
Lieferant: FLUOROCHEM
014251-100MG
014251-1G
FLUO014251-100MGEA
0
EUR
FLUO014251-100MG
FLUO014251-1G
3'-Methoxybiphenyl-4-carbonsäure
3'-Methoxybiphenyl-4-carbonsäure
Formel:
C₁₄H₁₂O₃ Molecular Weight: 228.25 g/mol |
MDL:
MFCD03424601 CAS-Nummer: 5783-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...