4'-Methoxybiphenyl-4-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
4'-Methoxy-[1,1'-biphenyl]-4-carboxylic acid, 4'-Methoxy[1,1'-biphenyl]-4-carboxylic acid
014252-100MG
014252-1G
FLUO014252-100MGEA
0
EUR
FLUO014252-100MG
FLUO014252-1G
4'-Methoxybiphenyl-4-carbonsäure
4'-Methoxybiphenyl-4-carbonsäure
Formel:
C₁₄H₁₂O₃ Molecular Weight: 228.25 g/mol |
MDL:
MFCD00185792 CAS-Nummer: 725-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...