4'-Methoxybiphenyl-4-carbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 4'-Methoxy-[1,1'-biphenyl]-4-carboxylic acid, 4'-Methoxy[1,1'-biphenyl]-4-carboxylic acid

8Y-0809.500 8Y-0809.1 8Y-0809.5 8Y-0809.10
BIOT8Y-0809.500EA 0 EUR
BIOT8Y-0809.500 BIOT8Y-0809.1 BIOT8Y-0809.5 BIOT8Y-0809.10
4'-Methoxybiphenyl-4-carbonsäure
4'-Methoxybiphenyl-4-carbonsäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
MDL: MFCD00185792
CAS-Nummer: 725-14-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR