4-(Benzyloxy)benzoesäure

Lieferant: FLUOROCHEM

002505-5G 002505-25G
FLUO002505-5GEA 72.9 EUR
FLUO002505-5G FLUO002505-25G
4-(Benzyloxy)benzoesäure
4-(Benzyloxy)benzoesäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
MDL: MFCD00016527
CAS-Nummer: 1486-51-7
EINECS: 216-066-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR