2'-Hydroxy-5'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2'-Hydroxy-5'-methoxy-[1,1'-biphenyl]-3-carboxylic acid

CA-4943-1G CA-4943-250M
COBBCA-4943-1GEA 862 EUR
COBBCA-4943-1G COBBCA-4943-250M
2'-Hydroxy-5'-methoxybiphenyl-3-carbonsäure
2'-Hydroxy-5'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₄
Molecular Weight: 244.25 g/mol
MDL: MFCD15143535
CAS-Nummer: 1215206-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR