4'-Hydroxy-3'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4941-1G CA-4941-250M
COBBCA-4941-1GEA 1180 EUR
COBBCA-4941-1G COBBCA-4941-250M
4'-Hydroxy-3'-methoxybiphenyl-3-carbonsäure
4'-Hydroxy-3'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₄
MDL: MFCD15143533
CAS-Nummer: 1215206-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR