2'-Hydroxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4818-5G CA-4818-1G
COBBCA-4818-5GEA 2460 EUR
COBBCA-4818-5G COBBCA-4818-1G
2'-Hydroxybiphenyl-3-carbonsäure
2'-Hydroxybiphenyl-3-carbonsäure
Formel: C₁₃H₁₀O₃
Molecular Weight: 214.22 g/mol
MDL: MFCD06802571
CAS-Nummer: 893736-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR