3'-Hydroxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3'-Hydroxy-[1,1'-biphenyl]-3-carboxylic acid

CA-4816-1G CA-4816-5G
COBBCA-4816-1GEA 452 EUR
COBBCA-4816-1G COBBCA-4816-5G
3'-Hydroxybiphenyl-3-carbonsäure
3'-Hydroxybiphenyl-3-carbonsäure
Formel: C₁₃H₁₀O₃
Molecular Weight: 214.22 g/mol
MDL: MFCD04117370
CAS-Nummer: 171047-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR