Biphenyl-2,2'-dicarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: [1,1'-Biphenyl]-2,2'-dicarboxylic acid, Diphenic acid, 2,2'-Biphenyldicarboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

126691-100G 126691-25G
SIAL126691-100GEA 116 EUR
SIAL126691-100G SIAL126691-25G
Biphenyl-2,2'-dicarbonsäure, Sigma-Aldrich®
Biphenyl-2,2'-dicarbonsäure
Formel: C₁₄H₁₀O₄
Molecular Weight: 242,23 g/mol
MDL: MFCD00002464
CAS-Nummer: 482-05-3
EINECS: 207-576-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR