Biphenyl-2,2'-dicarbonsäure

Lieferant: Thermo Scientific

Synonyme: [1,1'-Biphenyl]-2,2'-dicarboxylic acid, Diphenic acid, 2,2'-Biphenyldicarboxylic acid

SB00521.1 SB00521EA SB00521EE
MAYBSB00521.1EA 0 EUR
MAYBSB00521.1 MAYBSB00521EA MAYBSB00521EE
Biphenyl-2,2'-dicarbonsäure
Biphenyl-2,2'-dicarbonsäure
Formel: C₁₄H₁₀O₄
Molecular Weight: 242.23 g/mol
MDL: MFCD00002464
CAS-Nummer: 482-05-3
EINECS: 207-576-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR