Biphenyl-2,3'-dicarbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: [1,1'-Biphenyl]-2,3'-dicarboxylic acid

CA-4860-1G CA-4860-250M
COBBCA-4860-1GEA 404 EUR
COBBCA-4860-1G COBBCA-4860-250M
Biphenyl-2,3'-dicarbonsäure
Biphenyl-2,3'-dicarbonsäure
Formel: C₁₄H₁₀O₄
Molecular Weight: 242.23 g/mol
MDL: MFCD06203581
CAS-Nummer: 606-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR